Learn More
ANO2 Mouse anti-Human, Unconjugated, Clone: 2F12G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67638-1-IG-150UL
This item is not returnable.
View return policy
Description
The ANO2 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human ANO2. This antibody recognizes human, mouse antigen. The ANO2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ANO2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2F12G1 | |
Unconjugated | |
150 μL | |
57101 | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANO2 Mouse anti-Human, Unconjugated, Clone: 2F12G1, Proteintech > 150 μL
Spot an opportunity for improvement?Share a Content Correction