Learn More
ANO6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20784-1-AP-150UL
 View more versions of this product
Product Code. 16840355
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
May act as a calcium-activated chloride channel.
Specifications
Specifications
ANO6
Polyclonal
Unconjugated
ANO6
ANO6, anoctamin 6, TMEM16F, Transmembrane protein 16F
Rabbit
Antigen Affinity Chromatography
RUO
105722, 196527
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4KMQ2, Q6P9J9
Ano6
ANO6 Fusion Protein Ag13937
150 μL
Primary
Human, Mouse
Antibody
IgG
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ANO6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated