missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | AP2B1 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16874974
|
Proteintech
15690-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16864974
|
Proteintech
15690-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules.Specifications
AP2B1 | |
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P62944, P63010, Q9DBG3 | |
140670, 163, 71770 | |
AP2B1 Fusion Protein Ag8248 | |
Primary | |
-20°C | |
AP2B1 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ADTB2, AP 2 complex subunit beta, AP105B, AP2 BETA, AP2B1, Beta 2 adaptin, Beta adaptin, CLAPB1 | |
Ap2b1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AP2B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech