missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15690-1-AP-20UL
This item is not returnable.
View return policy
Description
Subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules.Specifications
AP2B1 | |
Polyclonal | |
Unconjugated | |
AP2B1 | |
ADTB2, AP 2 complex subunit beta, AP105B, AP2 BETA, AP2B1, Beta 2 adaptin, Beta adaptin, CLAPB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140670, 163, 71770 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62944, P63010, Q9DBG3 | |
Ap2b1 | |
AP2B1 Fusion Protein Ag8248 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AP2B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction