Learn More
Apolipoprotein AI Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14427-1-AP-20UL
 View more versions of this product
Product Code. 16823064
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This antibody was generated against the C-terminal region of human ApoA1.
Apolipoprotein A-1, also known as APOA1, is a human protein with a specific role in lipid metabolism. It binds to lipopolysaccharide or endotoxin, and has a major role in the anti-endotoxin function of HDL. The gene is mapped to 11q23. And it is a single polypeptide chain with 243 amino acid residues of known primary amino acid sequence. The ApoA-I protein promotes cholesterol efflux from tissues to the liver for excretion. It is a cofactor for lecithin cholesterolacyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. ApoA-I is also isolated as a prostacyclin (PGI2) stabilizing factor, and thus may have an anticlotting effect. Defects in the gene encoding it are associated with HDL deficiencies, including Tangier disease, and with systemic non-neuropathic amyloidosis. Additionally, ApoA-I overexpression promotes macrophage-specific reverse cholesterol transport.
Specifications
Specifications
Apolipoprotein AI
Polyclonal
Unconjugated
APOA1
Apo AI, ApoA I, APOA1, apolipoprotein A I, Apolipoprotein A1
Rabbit
Antigen Affinity Chromatography
RUO
11806, 335
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry
0.52 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02647, Q00623
Apoa1
Apolipoprotein AI Fusion Protein Ag5793
20 ÎĽL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Apolipoprotein AI Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated