Learn More
ARL2BP Mouse anti-Human, Unconjugated, Clone: 1G5C2, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ARL2BP |
---|---|
Clone | 1G5C2 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849875
|
Proteintech
67316-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16839875
|
Proteintech
67316-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ARL2BP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARL2BP. This antibody recognizes human antigen. The ARL2BP antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
ARL2BP | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
23568 | |
Primary |
1G5C2 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.