missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22025-1-AP-20UL
This item is not returnable.
View return policy
Description
May function as component of the Arp2/3 complex which is involved in regulation of actin polymerization and together with an activating nucleation-promoting factor (NPF) mediates the formation of branched actin networks.Specifications
ARPC5L | |
Polyclonal | |
Unconjugated | |
ARPC5L | |
ARC16 2, ARPC5L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296710, 74192, 81873 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A1L108, Q9BPX5, Q9D898 | |
Arpc5l | |
ARPC5L Fusion Protein Ag16881 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARPC5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction