Learn More
ASF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11011-1-AP-20UL
 View more versions of this product
Product Code. 16887783
  • 1645.54 SEK / 20µL
Description
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The encoded protein is the substrate of the tousled-like kinase family of cell cycle-regulated kinases, and may play a key role in modulating the nucleosome structure of chromatin by ensuring a constant supply of histones at sites of nucleosome assembly.
Specifications
Specifications
ASF1B
Polyclonal
Unconjugated
ASF1B
ASF1B, CCG1 interacting factor A II, CIA II, hAsf1, hAsf1b, hCIA II, Histone chaperone ASF1B
Rabbit
Antigen Affinity Chromatography
RUO
304648, 55723, 66929
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9DAP7, Q9NVP2
Asf1b
ASF1B Fusion Protein Ag1477
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ASF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated