missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16882-1-AP-150UL
This item is not returnable.
View return policy
Description
ATPase that plays a critical role in regulating the surface expression of AMPA receptors (AMPAR), thereby regulating synaptic plasticity and learning and memory. Required for NMDA-stimulated AMPAR internalization and inhibition of GRIA1 and GRIA2 recycling back to the plasma membrane; these activities are ATPase-dependent.Specifications
ATAD1 | |
Polyclonal | |
Unconjugated | |
ATAD1 | |
AFDC1, ATAD1, FNP001 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
309532, 67979, 84896 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q505J9, Q8NBU5, Q9D5T0 | |
ATAD1 | |
ATAD1 Fusion Protein Ag10343 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATAD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction