missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25316-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.Specifications
ATP6V1G2 | |
Polyclonal | |
Unconjugated | |
ATP6V1G2 | |
ATP6G, ATP6G2, ATP6V1G2, NG38, V ATPase 13 kDa subunit 2, V ATPase subunit G 2, VMA10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
368044, 534, 66237 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95670, Q9WTT4 | |
ATP6V1G2 | |
ATP6V1G2 Fusion Protein Ag18007 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATP6V1G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction