missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ATP6V1G2 |
---|---|
Concentration | 0.47 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883905
|
Proteintech
25316-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873905
|
Proteintech
25316-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.Specifications
ATP6V1G2 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O95670, Q9WTT4 | |
368044, 534, 66237 | |
ATP6V1G2 Fusion Protein Ag18007 | |
Primary | |
-20°C | |
ATP6V1G2 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATP6G, ATP6G2, ATP6V1G2, NG38, V ATPase 13 kDa subunit 2, V ATPase subunit G 2, VMA10 | |
ATP6V1G2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATP6V1G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech