Learn More
SymLine™ Automatic Switching Sets for Container
The ready-to-use automatic switching sets prevents the overflowing of disposal containers and the associated leakage of hazardous, toxic or explosive liquid waste.
Brand: SymLine™ 106752
27336.25 SEK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Description
- Coordinated system for monitoring the safe filling of disposal containers
- For connection to two disposal containers (not included in the set)
- Set consisting of Built-in Signalbox2, SwitchBox, ball valve and two signal cables
- Optionally available as an ATEX version with an additional switch amplifier for 2 channels
Specifications
CE | |
1 Built-in Signalbox2, EU/UK/US, 2-channel, 2 switch inputs, 2 switch outputs, 1x Switch amplifier set, 1x SwitchBox: control unit, 1x 3-way ball valve, 3x GL25 (m), electric, 1x 3-way ball valve mount, 2x signal cable | |
Disposal Container |
Automatic switching set for container (ATEX) | |
Potentially Explosive Atmospheres | |
Automatic Switching Set |
Product Content Correction
Product Title
SymLine™ Automatic Switching Sets for Container > Model: For potentially explosive atmospheres (ATEX)
Spot an opportunity for improvement?Share a Content Correction