missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ Automatic Switching Sets for Container
The ready-to-use automatic switching sets prevents the overflowing of disposal containers and the associated leakage of hazardous, toxic or explosive liquid waste.
20580.00 SEK - 28775.00 SEK
Specifications
Certifications/Compliance | CE |
---|---|
For Use With (Equipment) | Disposal Container |
Product Type | Automatic Switching Set |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | ||||||
18696478
|
SymLine™
106751 |
Non-explosive Atmospheres |
20580.00 SEK
1 set |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
18616488
|
SymLine™
106752 |
Potentially Explosive Atmospheres |
28775.00 SEK
1 set |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
Description
- Coordinated system for monitoring the safe filling of disposal containers
- For connection to two disposal containers (not included in the set)
- Set consisting of Built-in Signalbox2, SwitchBox, ball valve and two signal cables
- Optionally available as an ATEX version with an additional switch amplifier for 2 channels
Specifications
CE | |
Disposal Container | |
Automatic Switching Set |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SymLine™ Automatic Switching Sets for Container