missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG6 Mouse anti-Human, Mouse, Rat, Clone: 1B8D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66661-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein with two leucine zipper domains, a ribosomal protein S14 signature domain and a Sec39 like domain. The protein is thought to be involved in Golgi-to-ER transport. Mutations in this gene are associated with short stature, optic nerve atrophy, and Pelger-Huet anomaly. Involved in Golgi-to-endoplasmic reticulum (ER) retrograde tranport; the function is proposed to depend on its association in the NRZ complex which is believed to play a role in SNARE assembly at the ER.Specifications
BAG6 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46379, Q6MG49, Q9Z1R2 | |
BAG6 | |
BAG6 Fusion Protein Ag24525 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1B8D3 | |
Unconjugated | |
BAG6 | |
BAG 6, BAG6, BAT3, D6S52E, G3, HLA B associated transcript 3, Protein G3 | |
Mouse | |
Protein G | |
RUO | |
224727, 7917, 94342 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BAG6 Mouse anti-Human, Mouse, Rat, Clone: 1B8D3, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction