missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG6 Mouse anti-Human, Mouse, Rat, Clone: 1B8D3, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | BAG6 |
---|---|
Clone | 1B8D3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888745
|
Proteintech
66661-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878745
|
Proteintech
66661-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein with two leucine zipper domains, a ribosomal protein S14 signature domain and a Sec39 like domain. The protein is thought to be involved in Golgi-to-ER transport. Mutations in this gene are associated with short stature, optic nerve atrophy, and Pelger-Huet anomaly. Involved in Golgi-to-endoplasmic reticulum (ER) retrograde tranport; the function is proposed to depend on its association in the NRZ complex which is believed to play a role in SNARE assembly at the ER.Specifications
BAG6 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BAG 6, BAG6, BAT3, D6S52E, G3, HLA B associated transcript 3, Protein G3 | |
BAG6 | |
IgG1 | |
Protein G | |
Antibody |
1B8D3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Rat, Human, Mouse | |
P46379, Q6MG49, Q9Z1R2 | |
224727, 7917, 94342 | |
BAG6 Fusion Protein Ag24525 | |
Primary | |
-20°C | |
BAG6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BAG6 Mouse anti-Human, Mouse, Rat, Clone: 1B8D3, Proteintech