Learn More
Lenz Laborglasinstrumente™ Ball Members, Inch Range
Manufactured from DURAN™ tubing. Lenz Laborglasinstrumente™ Ball Members, Inch Range are perfect for mixing solutions in a centrifuge tube.
Brand:  Lenz Laborglasinstrumente™ 01214125
 View more versions of this product
Product Code. 10253951
  • 282.00 SEK / Each
Only null left
Estimated Shipment: 28-07-2025
Description
  • Acc. to DIN 12264 and in addition to standard
  • Made of DURAN™ tubing
Specifications
Specifications
Borosilicate Glass
120 mm
1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Ball Members, Inch Range > Overall Length (Metric): 120mm