missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12766-1-AP-150UL
This item is not returnable.
View return policy
Description
BBS4 contains tetratricopeptide repeats (TPR), similar to O-linked N-acetyglucosamine transferase. Mutations in the gene encoding this protein have been observed in patients with Bardet-Biedl syndrome type 4. BBS4 may play a role in pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation.Specifications
BBS4 | |
Polyclonal | |
Unconjugated | |
BBS4 | |
Bardet Biedl syndrome 4, BBS4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102774, 300754, 585 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C1Z7, Q96RK4 | |
BBS4 | |
BBS4 Fusion Protein Ag3430 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BBS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction