missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13458-1-AP-150UL
This item is not returnable.
View return policy
Specifications
BBS7 | |
Polyclonal | |
Unconjugated | |
BBS7 | |
Bardet Biedl syndrome 7, BBS2 like protein 1, BBS2L1, BBS7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361930, 55212, 71492 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IWZ6, Q8K2G4 | |
Bbs7 | |
BBS7 Fusion Protein Ag4314 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BBS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction