Learn More
BCAS3 Mouse anti-Human, Unconjugated, Clone: 1G7B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67085-1-IG-20UL
 View more versions of this product
Product Code. 16889505
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The BCAS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAS3. This antibody recognizes human, mouse, rat, pig antigen. The BCAS3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
BCAS3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
1G7B1
Unconjugated
20 μL
54828
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BCAS3 Mouse anti-Human, Unconjugated, Clone: 1G7B1, Proteintech > 20 μL