missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAS3 Mouse anti-Human, Unconjugated, Clone: 1G7B1, Proteintech
Mouse Monoclonal Antibody
1395.65 SEK - 3774.95 SEK
Specifications
Antigen | BCAS3 |
---|---|
Clone | 1G7B1 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
The BCAS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAS3. This antibody recognizes human, mouse, rat, pig antigen. The BCAS3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
BCAS3 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
54828 | |
Primary |
1G7B1 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BCAS3 Mouse anti-Human, Unconjugated, Clone: 1G7B1, Proteintech