Learn More
BCAS3 Mouse anti-Human, Unconjugated, Clone: 1G7B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67085-1-IG-150UL
Description
The BCAS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAS3. This antibody recognizes human, mouse, rat, pig antigen. The BCAS3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
BCAS3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
1G7B1 | |
Unconjugated | |
150 μL | |
54828 | |
IgG1 |