Learn More
BCAS3 Mouse anti-Human, Unconjugated, Clone: 1G7B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67085-1-IG-20UL
 View more versions of this product
Product Code. 16889505
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
The BCAS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAS3. This antibody recognizes human, mouse, rat, pig antigen. The BCAS3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
BCAS3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
1G7B1
Unconjugated
20 μL
54828
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BCAS3 Mouse anti-Human, Unconjugated, Clone: 1G7B1, Proteintech > 20 μL