Learn More
Bel-Art™ Pipet Vacuum Aspirator Collection System
Performs vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes.
Brand: Bel-Art™ F19917-0155
1147.90 SEK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Description
- Built-in pump quickly aspirates fluids from pipet tip, Pasteur pipet or serological pipet inserted into the collection tube
- Maintains low noise level (76 db at 1 meter) and minimal vibration
- Excellent for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
- Chemical and corrosion resistant polypropylene collection bottle holds 3800 mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
- A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
- Lightweight and compact design will not crowd workspace in a hood or at the bench
Specifications
Pipettes | |
Replacement Hydrophobic Filter | |
Bel-Art Vacuum Aspirator Collection System |
Plastic | |
3800 mL |
Promotions
-
Multiple Manufacturers
Genomics Powerhouse
Power your pipeline from PCR to sequencing.Promo Code: 24117Expires: 2025-08-24
New Offer Save Up to 92%