Learn More
Beta Arrestin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10171-1-AP-150UL
 View more versions of this product
Product Code. 16876373
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Beta-arrestin-2, or ARRB2, is an adaptor protein belonging to the arrestin family with an N- arrestin domain and a C- arrestin domain. ARRB2 is a regulatory protein involved in heterotrimeric G protein-coupled receptor desensitization and is known to regulate beta2-adrenergic receptor (beta2AR) function by enhancing beta2AR mediated nuclear translocation of ERK. ARRB2 binds to phosphorylated beta2ARs, thereby causing a significant impairment of their capacity to activate G(S) proteins. Along with AIP4, ARRB2 acts as an endosomal sorting molecule that mediates CXCR4 entry into a degradative pathway. It may also be involved in hormone-specific desensitization of TSH receptors. ARRB2 is a Ca2+ binding protein of the retinal outer segments and binds to P-rhodopsin and is also involved in synaptic transmission in photoreceptor cells. It is widely expressed in most of the tissues, especially neuronal tissues and spleen.
Specifications
Specifications
Beta Arrestin 2
Polyclonal
Unconjugated
ARRB2
ARB2, ARR2, ARRB2, Arrestin beta 2, arrestin, beta 2, BARR2, Beta arrestin 2
Rabbit
Antigen Affinity Chromatography
RUO
216869, 409
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P32121, Q91YI4
ARRB2
Beta Arrestin 2 Fusion Protein Ag0227
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Beta Arrestin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated