Learn More
beta Catenin Monoclonal Antibody (15B8), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-2567-95
Description
The 15B8 MAb reacts with human and mouse beta-catenin, one member of a family of catenins, which are intracellular proteins that interact with cadherins to mediate cellular adhesion. More specifically, beta-catenin binds to the cytoplasmic tail of E-cadherin. In addition, this molecule is a component of the canonical Wnt signaling pathway. In the absence of Wnt binding its receptor, beta-catenin is phosphorylated and resides in the cytoplasm where it is eventually targeted for degradation by ubiquitination. Upon Wnt binding, beta-catenin becomes dephosphorylated, translocates to the nucleus, and modulates gene expression in partnership with the transcription factors T cell factor (TCF) and lymphocyte enhancer binding factor (LEF). Expression of beta-catenin is found in a wide variety of non-immune and immune tissues, including thymocytes and T and B lymphocytes. The Wnt and beta-catenin signaling pathway has been demonstrated to play a crucial role in the development of T, B, and hematopoietic stem cells.
beta Catenin Monoclonal antibody specifically detects beta Catenin in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Specifications
beta Catenin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P35222, Q02248 | |
Mouse | |
10 mg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 Îş |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
15B8 | |
Unconjugated | |
CTNNB1 | |
armadillo; Bcatenin; B-catenin; beta catenin; Betacatenin; beta-catenin; beta-catenin 1; Bfc; C20orf33; catenin; catenin (cadherin associated protein), beta 1; catenin (cadherin associated protein), beta 1, 88kDa; catenin (cadherin-associated protein) beta 1; catenin (cadherin-associated protein), beta 1; catenin (cadherin-associated protein), beta 1, 88kDa; Catenin b 1; Catenin b1; catenin beta; catenin beta 1; catenin beta 1 L homeolog; Catenin beta1; catenin beta-1; Catenin Ăź 1; Catenin Ăź1; Catnb; CHBCAT; CTNB1; CTNNB; Ctnnb1; ctnnb1.L; ctnnb1-a; ctnnb1-b; dJ633O20.1; DKFZp686D02253; FLJ25606; FLJ37923; id:ibd2058; Mesc; MRD19; NAP; NYD-SP19; OK/SW-cl.35; OTTHUMP00000209289; P14L; PP8304; PRO2286; RP5-1118M15.1; wu:fb73e10; wu:fi81c06; wu:fk25h01; XELAEV_18031149mg; Ăź catenin; Ăźcatenin | |
Affinity chromatography | |
RUO | |
12387, 1499 | |
4° C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.