Learn More
Ansell BioClean™ Sterile Nitrile RABS/Isolator Sleeve GSL15NITPP27
Sterile Nitrile RABS/Isolator Sleeve is compatible with varied cuff systems and comes in either multiple sizes.
Marque: Ansell GSL36NITPP26
71174.78 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Assured quality: The BioClean™ Sterile Nitrile RABS/Isolator Sleeve GSL15NITPP26 is triple bagged, for assured integrity, and 100% visually inspected, ensuring high levels of quality
- Enhanced versatility: These nitrile protective sleeves are compatible with a wide range of cuff systems, extending protective coverage to the arm
- Specialized protection: In addition, they are autoclave compatible, offering wearers' arm protection in highly pressurized, high-temperature conditions
- Added benefits: These arm protection sleeves also feature extremely low levels of particles and offer excellent ESD properties
Spécification
EC 1935/2004, ISO 11137-1:2006, EN 420:2003 + A1:2009 | |
Class 10 (ISO 4) | |
Ambidextrous | |
Nitrile | |
12 | |
0.50 mm | |
12 in. | |
660 mm |
No | |
White | |
Yes | |
Isolator, RABS Sleeve | |
20 mil | |
Life Sciences Healthcare Electronics | |
26 in. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Ansell BioClean™ Sterile Nitrile RABS/Isolator Sleeve GSL15NITPP27 > Size: 12
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu