Learn More
Thermo Scientific Chemicals BIS-TRIS propane, 0.2M buffer soln., pH 9.0
CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol
978.00 SEK - 1490.00 SEK
Chemical Identifiers
CAS | 64431-96-5 |
---|---|
Molecular Formula | C11H26N2O6 |
Molecular Weight (g/mol) | 282.337 |
MDL Number | MFCD00004689 |
InChI Key | HHKZCCWKTZRCCL-UHFFFAOYSA-N |
PubChem CID | 125132 |
ChEBI | CHEBI:40947 |
IUPAC Name | 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(CNC(CO)(CO)CO)CNC(CO)(CO)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426099
|
Thermo Scientific Chemicals
J63943.AE |
100 mL |
978.00 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15436099
|
Thermo Scientific Chemicals
J63943.AK |
250 mL |
1490.00 SEK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64431-96-5 | |
282.337 | |
HHKZCCWKTZRCCL-UHFFFAOYSA-N | |
CHEBI:40947 | |
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO |
C11H26N2O6 | |
MFCD00004689 | |
125132 | |
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol |
Specifications
64431-96-5 | |
9.0 | |
C11H26N2O6 | |
HHKZCCWKTZRCCL-UHFFFAOYSA-N | |
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol | |
125132 | |
BIS-TRIS propane buffer soln. |
0.2 M | |
Liquid | |
MFCD00004689 | |
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO | |
282.337 | |
CHEBI:40947 |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.