Learn More
BRAND™ BLAUBRAND™ Bang Pattern Micro Burette
With interchangeable straight needle-valve stopcock with PTFE spindle (PTFE key in the intermediate stopcock)
4850.00 SEK - 5510.00 SEK
Specifications
No. per Pack 2
Class Class AS
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15281919
View Documents
BRAND™
24265
2 mL
5440.00 SEK
Pack of 2
Estimated Shipment: 11-06-2025
Add to basket
15291919
View Documents
BRAND™
24267
5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15201929
View Documents
BRAND™
24269
10 mL
5510.00 SEK
Pack of 2
Estimated Shipment: 11-06-2025
Add to basket
Description
Description
  • Class AS
  • Boro 3.3 Glass
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • Schellbach stripe
  • All BLAUBRAND™ burettes are delivered with batch number and enclosed one batch certificate per pack
  • Specifications
    Specifications
    2
    Class AS
    Documents
    Product Certifications
    Product Content Correction
    Product Title
    BRAND™ BLAUBRAND™ Bang Pattern Micro Burette