missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR1A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12702-1-AP-150UL
This item is not returnable.
View return policy
Description
The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.Specifications
BMPR1A | |
Polyclonal | |
Unconjugated | |
BMPR1A | |
10q23del, Activin receptor like kinase 3, ACVRLK3, ALK 3, ALK3, BMP type 1A receptor, BMPR 1A, BMPR1A, CD292, SKR5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
657 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36894 | |
BMPR1A | |
BMPR1A Fusion Protein Ag3410 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BMPR1A Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction