Learn More
Borate/NaoH Buffer, Chem Lab™ Product Code.: 11939272
Borate/NaoH Buffer, Chem Lab™
Product Code. 11939272
5000 mL, 5 L
Click to view available options
Quantity:
5000 mL
Unit Size:
5 L
Product Code. 11939272
Brand: Chem Lab CL03.0204.5500
Specifications
Melting Point 0°C
Boiling Point Boiling Point: 100°C
Color Colorless
pH 10
Physical Form Liquid
Quantity 5000 mL
Solution Type Buffer
Formulation pH 10 (±0.02/20°C) H3BO3-KCl-NaOH
Odor Odorless
Traceability to NIST Yes
Solubility Information Soluble in water
Show More Show Less
Product Content Correction
Product Title
Borate/NaoH Buffer, Chem Lab™ >