Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Removable Fritted Disc
Brand:  Kimble Chase™ 422230-5020
 View more versions of this product
Product Code. 11880522
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Ion exchange chromatography
20 mm
Type I
500 mm
With removable fritted disc
1 Each
Chromatography Column
150 mL
Borosilicate Glass
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kimble Chase™ Borosilicate Glass Chromatography Column with Removable Fritted Disc > Column Capacity: 150mL