Learn More
Trajan™ BP5 GC Capillary Column
A general purpose phase used for a wide range of applications
Brand:  Trajan™ 054184
 View more versions of this product
Product Code. 12985551
  • 13055.00 SEK / Each
Only null left
Estimated Shipment: 11-08-2025
Description
  • Excellent general purpose column
  • Low bleed
  • Non-polar
  • High temperature
  • Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
Specifications
320/340°C
5% Phenyl/95% Dimethyl Polysiloxane
Hydrocarbons Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons
0.25 ÎĽm
Forte™ BP5
Fused Silica
0.25 mm
G36, G41
-60°C to +320/340°C
60 m
Non polar
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Trajan™ BP5 GC Capillary Column > Film thickness: 0.25um; I.D.: 0.25mm; Length: 60m