Learn More
Trajan™ BP5 GC Capillary Column
A general purpose phase used for a wide range of applications
4740.00 SEK - 15070.00 SEK
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | ||||
10025290
|
Trajan™
054168 |
25 m | 0.22 mm | 0.25 ÎĽm |
8110.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
10605651
|
Trajan™
054167 |
12 m | 0.22 mm | 0.25 ÎĽm |
4740.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
10432531
|
Trajan™
054180 |
25 m | 0.32 mm | 0.25 ÎĽm |
8645.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
15258653
|
Trajan™
054178 |
60 m | 0.32 mm | 0.25 ÎĽm |
15070.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
15248653
|
Trajan™
054160 |
30 m | 0.53 mm | 3 ÎĽm |
10495.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
12985551
|
Trajan™
054184 |
60 m | 0.25 mm | 0.25 ÎĽm |
13055.00 SEK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||
15208663
|
Trajan™
054199 |
25 m | 0.53 mm | 1.5 ÎĽm |
5020.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
15278653
|
Trajan™
054188 |
60 m | 0.32 mm | 1 ÎĽm |
14370.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
15298653
|
Trajan™
054196 |
30 m | 0.53 mm | 5 ÎĽm |
9440.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
12995551
|
Trajan™
054216 |
30 m | 0.32 mm | 0.5 ÎĽm |
8620.00 SEK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
Description
- Excellent general purpose column
- Low bleed
- Non-polar
- High temperature
- Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
Non polar | |
Fused Silica |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Trajan™ BP5 GC Capillary Column