missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22236-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a transcriptional regulator that belongs to the BET (bromodomains and extra terminal domain) family of proteins. This protein associates with transcription complexes and with acetylated chromatin during mitosis, and it selectively binds to the acetylated lysine-12 residue of histone H4 via its two bromodomains. The gene maps to the major histocompatability complex (MHC) class II region on chromosome 6p21.3, but sequence comparison suggests that the protein is not involved in the immune response. This gene has been implicated in juvenile myoclonic epilepsy, a common form of epilepsy that becomes apparent in adolescence. Multiple alternatively spliced variants have been described for this gene, but the full-length nature of some of these variants has not been determined.Specifications
BRD2 | |
Polyclonal | |
Unconjugated | |
BRD2 | |
BRD2, bromodomain containing 2, D6S113E, FSH, FSRG1, KIAA9001, NAT, O27.1.1, RING3, RNF3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6046 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25440 | |
BRD2 | |
BRD2 Fusion Protein Ag17589 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
BRD2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur