missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | BRD2 |
---|---|
Concentration | 0.81 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891775
|
Proteintech
22236-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881775
|
Proteintech
22236-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a transcriptional regulator that belongs to the BET (bromodomains and extra terminal domain) family of proteins. This protein associates with transcription complexes and with acetylated chromatin during mitosis, and it selectively binds to the acetylated lysine-12 residue of histone H4 via its two bromodomains. The gene maps to the major histocompatability complex (MHC) class II region on chromosome 6p21.3, but sequence comparison suggests that the protein is not involved in the immune response. This gene has been implicated in juvenile myoclonic epilepsy, a common form of epilepsy that becomes apparent in adolescence. Multiple alternatively spliced variants have been described for this gene, but the full-length nature of some of these variants has not been determined.Specifications
BRD2 | |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
P25440 | |
6046 | |
BRD2 Fusion Protein Ag17589 | |
Primary | |
-20°C | |
BRD2 |
0.81 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BRD2, bromodomain containing 2, D6S113E, FSH, FSRG1, KIAA9001, NAT, O27.1.1, RING3, RNF3 | |
BRD2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRD2 Rabbit anti-Human, Polyclonal, Proteintech