missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTF3L3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19753-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody has no cross reaction to BTF3.
This locus defines a putative member of the BTF3 family of transcription factors and is thought to represent a pseudogene.Specifications
BTF3L3 | |
Polyclonal | |
Unconjugated | |
BTF3P13 | |
BTF3P13 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BTF3L3, HUMBTFD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132556 | |
-20°C | |
Liquid |