missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTF3L3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | BTF3L3 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879234
|
Proteintech
19753-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869234
|
Proteintech
19753-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody has no cross reaction to BTF3.
This locus defines a putative member of the BTF3 family of transcription factors and is thought to represent a pseudogene.Specifications
BTF3L3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
BTF3L3, HUMBTFD | |
BTF3P13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
132556 | |
Peptide | |
Primary | |
-20°C | |
BTF3P13 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTF3L3 Rabbit anti-Human, Polyclonal, Proteintech