Learn More
BTG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20514-1-AP-20UL
 View more versions of this product
Product Code. 16890055
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Shows marked antiproliferative activity, being able to induce G(1) arrest.
Specifications
Specifications
BTG4
Polyclonal
Unconjugated
BTG4
B cell translocation gene 4, BTG family member 4, BTG4, PC3B, Protein BTG4, Protein PC3b
Rabbit
Antigen Affinity Chromatography
RUO
315650, 54766, 56057
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70552, Q9NY30
Btg4
BTG4 Fusion Protein Ag14339
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated