missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN3A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25221-1-AP-20UL
This item is not returnable.
View return policy
Description
The butyrophilin (BTN) genes are a group of major histocompatibility complex (MHC)-associated genes that encode type I membrane proteins with 2 extracellular immunoglobulin (Ig) domains and an intracellular B30.2 (PRYSPRY) domain. Three subfamilies of human BTN genes are located in the MHC class I region: the single-copy BTN1A1 gene and the BTN2 and BTN3 genes, which have undergone tandem duplication, resulting in 3 copies of each.Specifications
BTN3A1 | |
Polyclonal | |
Unconjugated | |
BTN3A1 | |
BT3.1, BTF5, BTN3A1, CD277 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11119 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00481 | |
BTN3A1 | |
BTN3A1 Fusion Protein Ag18414 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTN3A1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction