missing translation for 'onlineSavingsMsg'
Learn More

C15orf58 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25447-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884025

  • 1355.00 SEK / 20µL
Estimated Shipment: 30-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specific and highly efficient GDP-D-glucose phosphorylase regulating the levels of GDP-D-glucose in cells.
Specifications

Specifications

C15orf58
Polyclonal
Unconjugated
GDPGP1
C15orf58, UPF0580 protein C15orf58
Rabbit
Antigen Affinity Chromatography
RUO
269952, 390637
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3TLS3, Q6ZNW5
GDPGP1
C15orf58 Fusion Protein Ag20215
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers