Learn More
C1orf187 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26342-1-AP-20UL
 View more versions of this product
Product Code. 16884835
  • 1645.54 SEK / 20µL
Description
The C1orf187 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human C1orf187. This antibody recognizes human, mouse antigen. The C1orf187 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
C1orf187
Polyclonal
Unconjugated
20 μL
374946
IgG
Western Blot, ELISA
200 μg/mL
Rabbit
Primary
Human, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
C1orf187 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL