Learn More
C1orf187 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | C1orf187 |
---|---|
Concentration | 200 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884835
|
Proteintech
26342-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874835
|
Proteintech
26342-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The C1orf187 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human C1orf187. This antibody recognizes human, mouse antigen. The C1orf187 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
C1orf187 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG |
200 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
374946 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.