missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf86 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | C1orf86 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884665
|
Proteintech
26138-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874665
|
Proteintech
26138-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C1orf86 is a component of the Fanconi anemia (FA) complex required to recruit the FA complex to DNA interstrand cross-links (ICLs) and promote ICLs repair. Following DNA damage recognizes and binds ′Lys-63′-linked ubiquitin generated by RNF8 at ICLs and recruits other components of the FA complex. It promotes translesion synthesis via interaction with REV1.Specifications
C1orf86 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6NZ36 | |
199990 | |
C1orf86 Fusion Protein Ag23534 | |
Primary | |
-20°C | |
FAAP20 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C1orf86, FAAP20, FP7162 | |
FAAP20 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C1orf86 Rabbit anti-Human, Polyclonal, Proteintech