Learn More
C9orf72 Mouse, anti-Human, Unconjugated, Clone: 2F9A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67824-1-IG-20UL
 View more versions of this product
Product Code. 17231476
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
C9ORF72 has a domain whith polymorphic hexanucleotide repeat (GGGGCC). The C9ORF72-hexanucleotide repeat expansions have been recently identified as genetic markers in amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD). The C9ORF72 repeat expansions may indicate a worse prognosis in ALS. Human C9ORF72 has some isoforms with MW 54-60 kDa and 25-30 kDa. Mouse C9orf72 has some isoforms with MW 50-60 kDa and 35 kDa.
Specifications
Specifications
C9orf72
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
2F9A2
Unconjugated
20 μL
203228
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C9orf72 Mouse, anti-Human, Unconjugated, Clone: 2F9A2, Proteintech > 20 μL