Learn More
C9orf72 Mouse, anti-Human, Unconjugated, Clone: 2F9A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67824-1-IG-150UL
 View more versions of this product
Product Code. 17221476
  • 4652.90 SEK / 150µL
Description
C9ORF72 has a domain whith polymorphic hexanucleotide repeat (GGGGCC). The C9ORF72-hexanucleotide repeat expansions have been recently identified as genetic markers in amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD). The C9ORF72 repeat expansions may indicate a worse prognosis in ALS. Human C9ORF72 has some isoforms with MW 54-60 kDa and 25-30 kDa. Mouse C9orf72 has some isoforms with MW 50-60 kDa and 35 kDa.
Specifications
Specifications
C9orf72
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
2F9A2
Unconjugated
150 ÎĽL
203228
Liquid
Certificates
Promotions
Promotions
Product Content Correction
Product Title
C9orf72 Mouse, anti-Human, Unconjugated, Clone: 2F9A2, Proteintech > 150 ÎĽL