Learn More
C9orf72 Mouse, anti-Human, Unconjugated, Clone: 2F9A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67824-1-IG-150UL
Description
C9ORF72 has a domain whith polymorphic hexanucleotide repeat (GGGGCC). The C9ORF72-hexanucleotide repeat expansions have been recently identified as genetic markers in amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD). The C9ORF72 repeat expansions may indicate a worse prognosis in ALS. Human C9ORF72 has some isoforms with MW 54-60 kDa and 25-30 kDa. Mouse C9orf72 has some isoforms with MW 50-60 kDa and 35 kDa.
Specifications
C9orf72 | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
2F9A2 | |
Unconjugated | |
150 ÎĽL | |
203228 | |