missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALHM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19931-1-AP-150UL
This item is not returnable.
View return policy
Description
CALHM2, also named as FAM26B, is a pore-forming subunit of a voltage-gated ion channel. It is a critical ATP-releasing channel that modulates neural activity and as a potential risk factor of depression. There are three isoforms of CALHM2 with calculated MW 36 kDa, 24 kDa and 22 kDa. It is about 32-36 kDa in western blot with membrane protein extracted lysate.Specifications
CALHM2 | |
Polyclonal | |
Unconjugated | |
CALHM2 | |
CALHM2, FAM26B, Protein FAM26B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294019, 51063, 72691 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5RJQ8, Q8VEC4, Q9HA72 | |
CALHM2 | |
CALHM2 Fusion Protein Ag13909 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CALHM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction