missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZA1 Mouse anti-Human, Mouse, Rat, Clone: 10H1F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66066-1-IG-150UL
This item is not returnable.
View return policy
Description
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.Specifications
CAPZA1 | |
Monoclonal | |
0.85 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2GUZ5, P47753, P52907 | |
CAPZA1 | |
CAPZA1 Fusion Protein Ag18841 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
10H1F9 | |
Unconjugated | |
CAPZA1 | |
CAPPA1, CAPZ, CapZ alpha 1, CAPZA1, CAZ1 | |
Mouse | |
Protein G | |
RUO | |
12340, 691149, 829 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CAPZA1 Mouse anti-Human, Mouse, Rat, Clone: 10H1F9, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction