missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATSPERB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26633-1-AP-20UL
This item is not returnable.
View return policy
Description
CATSPERB, also named as C14orf161 and CatSper-beta, is involved in sperm cell hyperactivation via its association with CATSPER1. CATSPERB has two isoforms with MW 127 kDa and 72 kDa.Specifications
CATSPERB | |
Polyclonal | |
Unconjugated | |
CATSPERB | |
C14orf161, CatSper beta, CatSper(beta), CATSPERB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79820 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H7T0 | |
CATSPERB | |
CATSPERB Fusion Protein Ag23393 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CATSPERB Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction