Learn More
CCDC56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20427-1-AP-20UL
 View more versions of this product
Product Code. 16879964
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Specifications
CCDC56
Polyclonal
Unconjugated
COA3
CCDC56, HSPC009
Rabbit
Antigen Affinity Chromatography
RUO
28958, 498000, 52469
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D2R6, Q9Y2R0
COA3
CCDC56 Fusion Protein Ag14260
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCDC56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated